siRNA Details
Gene Symbol: Tsc22d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081229.1 | NP_001074698.1 | 4 | 2103 | ||
GenBank mRNA | AK007760.1 | - | 3 | 446 | ||
AK156074.1 | - | 2 | 87 | |||
BC058221.1 | - | 3 | 299 | |||
BC117998.1 | - | 4 | 1832 | 40095084 | ||
EU004151.1 | - | 4 | 488 |
