siRNA Details
Gene Symbol: Cnot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037846.3 | NP_001032935.2 | 14 | 1476 | ||
NM_001037847.2 | NP_001032936.2 | 14 | 1406 | |||
NM_028082.2 | NP_082358.2 | 13 | 1353 | |||
GenBank mRNA | AK011231.1 | - | 12 | 1276 | ||
AK078121.1 | - | 14 | 1466 | |||
AK149767.1 | - | 14 | 1450 | |||
AK167834.1 | - | 14 | 1454 | |||
BC043133.1 | - | 14 | 1377 | |||
BC063105.1 | - | 15 | 1619 | 5003026 | ||
BC065171.1 | - | 14 | 1406 | 6415034 | ||
BC090624.1 | - | 12 | 1282 | 6815664 |

More Information
Other Species Targeted

Gene Ontology Categories:
