siRNA Details
Gene Symbol: Cnot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037846.3 | NP_001032935.2 | 15 | 1577 | ||
NM_001037847.2 | NP_001032936.2 | 15 | 1507 | |||
NM_028082.2 | NP_082358.2 | 14 | 1454 | |||
GenBank mRNA | AK011231.1 | - | 13 | 1377 | ||
AK078121.1 | - | 15 | 1567 | |||
AK149767.1 | - | 15 | 1551 | |||
AK167834.1 | - | 15 | 1555 | |||
BC043133.1 | - | 15 | 1478 | |||
BC063105.1 | - | 16 | 1720 | 5003026 | ||
BC065171.1 | - | 15 | 1507 | 6415034 | ||
BC090624.1 | - | 13 | 1383 | 6815664 |

More Information
Other Species Targeted

Gene Ontology Categories:
