siRNA Details
Gene Symbol: Tmem158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002267.2 | NP_001002267.2 | 1 | 364 | ||
GenBank mRNA | AB114272.1 | - | 1 | 120 | ||
AK009669.1 | - | 1 | 340 | |||
BC139427.1 | - | 1 | 140 | 8862449 | ||
BC139428.1 | - | 1 | 140 | 8862450 |
