siRNA Details
Gene Symbol: Hsdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301367.1 | NP_001288296.1 | 10 | 1218 | ||
NM_024255.3 | NP_077217.2 | 9 | 1120 | |||
GenBank mRNA | AJ293845.1 | - | 10 | 1142 | ||
AK011910.1 | - | 9 | 1068 | |||
AK085899.1 | - | 9 | 1089 | |||
AK193150.1 | - | 2 | 51 | |||
AY725196.1 | - | 9 | 977 | |||
BC002211.1 | - | 9 | 993 | 3490303 |
