siRNA Details
Gene Symbol: Hsdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301367.1 | NP_001288296.1 | 4, 5 | 570 | ||
NM_024255.3 | NP_077217.2 | 3, 4 | 472 | |||
GenBank mRNA | AJ293845.1 | - | 4, 5 | 493 | ||
AK011910.1 | - | 3, 4 | 430 | |||
AK085899.1 | - | 3, 4 | 441 | |||
AK136773.1 | - | 3, 4 | 439 | |||
AK144948.1 | - | 3, 4 | 388 | |||
AK145230.1 | - | 3, 4 | 332 | |||
AK202946.1 | - | 2, 3 | 187 | |||
AK205169.1 | - | 2, 3 | 187 | |||
AK211849.1 | - | 2, 3 | 187 | |||
AK213102.1 | - | 2, 3 | 179 | |||
AK215396.1 | - | 2, 3 | 187 | |||
AY725196.1 | - | 3, 4 | 329 | |||
BC002211.1 | - | 3, 4 | 345 | 3490303 |
