siRNA Details
Gene Symbol: Hsdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301367.1 | NP_001288296.1 | 9 | 1103 | ||
NM_024255.3 | NP_077217.2 | 8 | 1005 | |||
GenBank mRNA | AJ293845.1 | - | 9 | 1027 | ||
AK085899.1 | - | 8 | 974 | |||
AK136773.1 | - | 8 | 972 | |||
AK144948.1 | - | 8 | 921 | |||
AK145230.1 | - | 8 | 865 | |||
AY725196.1 | - | 8 | 862 | |||
BC002211.1 | - | 8 | 878 | 3490303 |
