siRNA Details
Gene Symbol: Shisa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174086.1 | NP_001167557.1 | 4 | 1511 | ||
NM_028277.2 | NP_082553.2 | 5 | 1559 | |||
GenBank mRNA | AK049860.1 | - | 5 | 1085 | ||
BC030884.1 | - | 4 | 800 | |||
GU479981.1 | - | 5 | 1088 | |||
GU479982.1 | - | 4 | 1040 |
