siRNA Details
Gene Symbol: Zfp839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199785.1 | NP_001186714.1 | 7 | 2347 | ||
NM_028365.2 | NP_082641.1 | 8 | 2575 | |||
GenBank mRNA | AK013356.1 | - | 7 | 2320 | ||
BC150732.1 | - | 8 | 2575 | 9087643 |
