siRNA Details
Gene Symbol: Setd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028385.1 | NP_082661.1 | 6 | 663 | ||
GenBank mRNA | AK013649.1 | - | 2 | 593 | ||
AK038052.1 | - | 1 | 1025 | |||
AK122551.1 | - | 6 | 663 | |||
AK164036.1 | - | 1 | 2568 | |||
BC056502.1 | - | 8 | 1199 | |||
BC083184.1 | - | 5 | 544 | 30536831 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106614, XM_006237014, XM_008763187, XM_006237012, XM_008763189, and others.
Gene Ontology Categories:
