siRNA Details
Gene Symbol: 3110009E18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172074.1 | NP_001165545.1 | 3, 4 | 811 | ||
NM_028439.2 | NP_082715.1 | 3, 4 | 218 | |||
GenBank mRNA | AK014026.1 | - | 3, 4 | 812 | ||
BC028767.1 | - | 3, 4 | 210 | 1264217 |
