siRNA Details
Gene Symbol: 3110009E18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028439.2 | NP_082715.1 | 4, 5 | 257 | ||
GenBank mRNA | BC028767.1 | - | 4, 5 | 249 | 1264217 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 3110009E18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028439.2 | NP_082715.1 | 4, 5 | 257 | ||
GenBank mRNA | BC028767.1 | - | 4, 5 | 249 | 1264217 |