siRNA Details
Gene Symbol: Mrgbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028479.1 | NP_082755.1 | 2 | 538 | ||
GenBank mRNA | AK005551.1 | - | 1 | 178 | ||
AK038431.1 | - | 1 | 184 | |||
AK052575.1 | - | 1 | 193 | |||
BC042508.1 | - | 1 | 152 | |||
BC054431.1 | - | 1 | 125 | |||
BC085320.1 | - | 1 | 199 | |||
BC110993.1 | - | 1 | 147 | 5363496 | ||
BC116732.1 | - | 2 | 538 | 40126051 | ||
BC116734.1 | - | 2 | 538 | 40126053 |
