siRNA Details
Gene Symbol: Ankrd45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028664.1 | NP_082940.1 | 2 | 467 | ||
GenBank mRNA | AK016757.1 | - | 1 | 386 | ||
BC049713.1 | - | 2 | 439 | 6741553 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK144074.1 | - | 1852 | Chr. - - [-] on Build |
