siRNA Details
Gene Symbol: Jakmip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028708.2 | NP_082984.1 | 490 | |||
GenBank mRNA | AK018189.2 | - | 492 | |||
AK220482.1 | - | 470 | ||||
BC119557.1 | - | 380 | ||||
BC119558.1 | - | 377 |
