siRNA Details
Gene Symbol: Pex26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014598.1 | - | 3 | 818 | ||
AK040411.1 | - | 808 | ||||
AK042663.1 | - | 3 | 818 | |||
AK044186.1 | - | 3 | 770 | |||
AK087637.1 | - | 3 | 1040 | |||
AK147925.1 | - | 3 | 810 | |||
BC019144.1 | - | 3 | 801 | 5050939 | ||
BC058694.1 | - | 3 | 791 | 6811976 |
