siRNA Details
Gene Symbol: Pex26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014598.1 | - | 5 | 1251 | ||
AK040411.1 | - | 1241 | ||||
AK042663.1 | - | 5 | 1248 | |||
AK044186.1 | - | 5 | 1200 | |||
AK087637.1 | - | 5 | 1473 | |||
AK145961.1 | - | 4 | 956 | |||
AK147925.1 | - | 5 | 1243 | |||
BC019144.1 | - | 5 | 1234 | 5050939 | ||
BC058694.1 | - | 5 | 1224 | 6811976 |
