siRNA Details
Gene Symbol: Fbxl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175206.4 | NP_780415.1 | 2 | 558 | ||
GenBank mRNA | AK003404.1 | - | 2 | 574 | ||
AK052175.1 | - | 2 | 544 | |||
AK084845.1 | - | 2 | 474 | |||
AK085980.1 | - | 2 | 501 | |||
BC100535.1 | - | 2 | 554 | 30138645 |
