siRNA Details
Gene Symbol: Cmip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163262.1 | NP_001156734.1 | 19 | 2203 | ||
NM_028941.1 | NP_083217.1 | 19 | 2026 | |||
GenBank mRNA | AK016712.1 | - | 19 | 2027 | ||
AK165728.1 | - | 8 | 758 | |||
AK173234.1 | - | 13 | 4453 | |||
BC028834.1 | - | 17 | 1695 | |||
BC158118.1 | - | 20 | 2080 | 9088191 |
