siRNA Details
Gene Symbol: Cmip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163262.1 | NP_001156734.1 | 7 | 890 | ||
NM_028941.1 | NP_083217.1 | 7 | 713 | |||
GenBank mRNA | AK016712.1 | - | 7 | 714 | ||
AK085367.1 | - | 7 | 793 | |||
AK173234.1 | - | 1 | 3140 | |||
BC028834.1 | - | 5 | 382 | |||
BC158118.1 | - | 8 | 767 | 9088191 | ||
CA552751.1 | - | 4 | 281 |
