siRNA Details
Gene Symbol: Mgme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289630.1 | NP_001276559.1 | 2, 3 | 998 | ||
NM_001289631.1 | NP_001276560.1 | 2, 3 | 669 | |||
NM_028984.5 | NP_083260.1 | 2, 3 | 727 | |||
GenBank mRNA | AK018387.1 | - | 2, 3 | 656 | ||
AK031535.1 | - | 2, 3 | 597 | |||
AK043140.1 | - | 2, 3 | 666 | |||
AK049565.1 | - | 2, 3 | 868 | |||
AK159286.1 | - | 2, 3 | 649 | |||
AK167919.1 | - | 2, 3 | 656 | |||
BC063088.1 | - | 2, 3 | 642 | 6329248 |
