siRNA Details
Gene Symbol: Tbc1d30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029057.1 | NP_083333.1 | 11 | 1771 | ||
GenBank mRNA | AK015700.1 | - | 5 | 504 | ||
AK044521.1 | - | 11 | 1578 | |||
AK049989.1 | - | 11 | 1604 | |||
AK173079.1 | - | 11 | 1534 | |||
BC059081.1 | - | 3 | 279 | |||
BC072607.1 | - | 11 | 1597 | |||
BC079587.1 | - | 8 | 1082 | |||
BC138285.1 | - | 11 | 1978 | 8861304 | ||
BC138286.1 | - | 11 | 1978 | 8861305 |
