siRNA Details
Gene Symbol: Rhoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081105.1 | NP_001074574.1 | 3 | 1132 | ||
GenBank mRNA | AK017885.1 | - | 3 | 1118 | ||
AK137909.1 | - | 1 | 563 | |||
AK138090.1 | - | 4 | 1259 | |||
AK169618.1 | - | 3 | 1093 | |||
BC040409.1 | - | 1 | 507 | |||
BC094937.1 | - | 3 | 871 | 30075117 |
