siRNA Details
Gene Symbol: Meikin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB987828.1 | - | 12 | 1096 | ||
AK015073.1 | - | 12 | 1115 | |||
BC107368.1 | - | 11 | 1204 | 40056257 | ||
BC107369.1 | - | 12 | 1113 |
