siRNA Details
Gene Symbol: Meikin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB987828.1 | - | 13 | 1169 | ||
AK015073.1 | - | 13 | 1188 | |||
BC107368.1 | - | 12 | 1277 | 40056257 | ||
BC107369.1 | - | 13 | 1186 |
