siRNA Details
Gene Symbol: 1700009N14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081095.1 | NP_001074564.1 | 2 | 871 | ||
GenBank mRNA | AK005799.1 | - | 2 | 876 | ||
BC050973.1 | - | 2 | 647 | |||
BC116915.1 | - | 2 | 720 | 40126234 | ||
BC116917.1 | - | 2 | 720 | 40126236 |
