siRNA Details
Gene Symbol: 9130401M01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029418.4 | NP_083694.1 | 6 | 1197 | ||
GenBank mRNA | AK018655.1 | - | 6 | 1121 | ||
AK047927.1 | - | 6 | 1164 | |||
AK089715.1 | - | 1121 | ||||
AK167012.1 | - | 6 | 1219 | |||
AY053393.1 | - | 6 | 1220 | |||
BC022163.1 | - | 1 | 1901 | |||
BC050895.1 | - | 6 | 1197 | 5714794 |
