siRNA Details
Gene Symbol: Lca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027448.2 | NP_081724.1 | 6 | 1419 | ||
NM_029434.3 | NP_083710.2 | 6 | 1315 | |||
GenBank mRNA | AK015260.1 | - | 6 | 1287 | ||
BC049101.1 | - | 6 | 1347 | 5702365 | ||
BC052060.1 | - | 6 | 1347 | 5702365 |
