siRNA Details
Gene Symbol: 4930584F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005600.1 | - | 4 | 544 | ||
AK015867.1 | - | 2 | 309 | |||
AK016349.1 | - | 3 | 356 | |||
BC049642.1 | - | 3 | 586 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930584F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005600.1 | - | 4 | 544 | ||
AK015867.1 | - | 2 | 309 | |||
AK016349.1 | - | 3 | 356 | |||
BC049642.1 | - | 3 | 586 |