siRNA Details
Gene Symbol: Adhfe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175236.4 | NP_780445.1 | 6 | 563 | ||
GenBank mRNA | AK018241.1 | - | 6 | 537 | ||
AK038853.1 | - | 6 | 530 | |||
AK046780.1 | - | 6 | 489 | |||
AK050178.1 | - | 5 | 487 | |||
AK084104.1 | - | 491 | ||||
BC026584.1 | - | 6 | 521 | 4972323 | ||
CA467848.1 | - | 6 | 602 |
