siRNA Details
Gene Symbol: Slc24a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110240.1 | NP_001103710.1 | 1 | 796 | ||
NM_172426.2 | NP_766014.1 | 2 | 1021 | |||
GenBank mRNA | AK034424.1 | - | 2 | 905 | ||
AK038634.1 | - | 2 | 905 | |||
AK038800.1 | - | 2 | 949 | |||
AK083127.1 | - | 2 | 905 | |||
AK136058.1 | - | 2 | 950 | |||
BC115867.1 | - | 1 | 798 | 40081382 | ||
BC116637.1 | - | 1 | 798 | 40081381 |
