siRNA Details
Gene Symbol: Abcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029600.3 | NP_083876.3 | 10 | 1349 | ||
GenBank mRNA | AF534127.1 | - | 10 | 1349 | ||
AF534128.1 | - | 10 | 1349 | |||
AF534129.1 | - | 10 | 1349 | |||
AK149551.1 | - | 10 | 1349 | |||
AK155664.1 | - | 10 | 1324 | |||
AK156615.1 | - | 10 | 1579 | |||
AK172420.1 | - | 10 | 1579 | |||
AY795569.1 | - | 10 | 1349 | |||
AY841885.1 | - | 10 | 1330 | |||
BC046560.1 | - | 7 | 853 | |||
BC048825.1 | - | 10 | 1281 | |||
BC150788.1 | - | 10 | 1340 | 9087699 |
