siRNA Details
Gene Symbol: Mib2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256107.1 | NP_001243036.1 | 17 | 3132 | ||
NM_001256108.2 | NP_001243037.2 | 17 | 2952 | |||
NM_145124.3 | NP_660106.2 | 16 | 3036 | |||
GenBank mRNA | AK008671.1 | - | 3 | 899 | ||
AK084791.1 | - | 8 | 1706 | |||
AK147940.1 | - | 17 | 2898 | |||
AK159611.1 | - | 17 | 2849 | |||
AY974090.1 | - | 15 | 2333 | |||
BC058086.1 | - | 14 | 2620 | |||
BC141133.1 | - | 14 | 2425 | 9055745 | ||
BC145441.1 | - | 15 | 2521 |
