siRNA Details
Gene Symbol: Ctrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033875.2 | NP_001029047.1 | 4 | 412 | ||
GenBank mRNA | AK007772.1 | - | 3 | 240 | ||
BC103715.1 | - | 4 | 410 | 6430373 | ||
BC115516.1 | - | 4 | 412 | 40101127 | ||
BC115517.1 | - | 4 | 412 | 40101128 |
