siRNA Details
Gene Symbol: 1700039E15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033176.1 | NP_001028348.1 | 6 | 941 | ||
GenBank mRNA | AK133083.1 | - | 6 | 941 | ||
BC100502.1 | - | 2 | 743 | |||
BC147591.1 | - | 6 | 1038 | 9056581 | ||
BC147593.1 | - | 6 | 1038 | 9056583 | ||
BC172112.1 | - | 6 | 979 |
