siRNA Details
Gene Symbol: Bri3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029752.2 | NP_084028.1 | 3 | 793 | ||
GenBank mRNA | AK010810.1 | - | 3 | 736 | ||
AK043288.1 | - | 3 | 735 | |||
AK044565.1 | - | 3 | 681 | |||
AK143538.1 | - | 3 | 752 | |||
BC039941.1 | - | 3 | 751 | 4011957 |
