siRNA Details
Gene Symbol: Mnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029797.3 | NP_084073.2 | 7, 8 | 627 | ||
GenBank mRNA | AK011664.1 | - | 7, 8 | 537 | ||
BC027741.1 | - | 7, 8 | 554 | 5035682 | ||
BG070803.2 | - | 1, 2 | 244 | |||
BY710901.1 | - | 7, 8 | 537 |
Gene Symbol: Mnd1-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156325.1 | - | 1 | 2327 |
