siRNA Details
Gene Symbol: 2700049A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163378.1 | NP_001156850.1 | 25 | 4824 | ||
NM_029818.1 | NP_084094.1 | 25 | 4824 | |||
GenBank mRNA | AK012399.1 | - | 10 | 1504 | ||
AK172976.1 | - | 25 | 4474 | |||
BC043104.1 | - | 25 | 4824 | |||
BC150744.1 | - | 25 | 4192 | 9087655 |
