siRNA Details
Gene Symbol: Wdr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029896.1 | NP_084172.1 | 6 | 908 | ||
GenBank mRNA | AK035109.1 | - | 6 | 910 | ||
AK037620.1 | - | 6 | 915 | |||
AK149692.1 | - | 6 | 909 | |||
AK159240.1 | - | 6 | 908 | |||
BC019115.1 | - | 5 | 434 | |||
BC031502.1 | - | 6 | 598 | |||
BC094419.1 | - | 6 | 871 | |||
BC157943.1 | - | 831 | 9007278 | |||
BC157945.1 | - | 832 | 9007280 | |||
BC158054.1 | - | 832 | 9007356 | |||
EF011615.1 | - | 6 | 597 |
