siRNA Details
Gene Symbol: Vps33a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029929.3 | NP_084205.3 | 3 | 299 | ||
GenBank mRNA | AF439858.1 | - | 3 | 283 | ||
AK019463.1 | - | 3 | 283 | |||
AK041982.1 | - | 3 | 295 | |||
AK140553.1 | - | 3 | 288 | |||
AK150282.1 | - | 3 | 291 | |||
AK150550.1 | - | 3 | 291 | |||
AK179865.1 | - | 50 | ||||
AK187752.1 | - | 50 | ||||
BB852750.1 | - | 3 | 323 | |||
BC046417.1 | - | 3 | 290 | 4501207 |
