siRNA Details
Gene Symbol: C330007P06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029951.1 | NP_084227.1 | 2, 3 | 366 | ||
GenBank mRNA | AK021190.1 | - | 2, 3 | 359 | ||
AK032304.1 | - | 2, 3 | 281 | |||
AK034943.1 | - | 2, 3 | 307 | |||
AK035825.1 | - | 2, 3 | 355 | |||
AK036910.1 | - | 2, 3 | 289 | |||
AK044209.1 | - | 2, 3 | 303 | |||
AK084447.1 | - | 2, 3 | 296 | |||
AK086009.1 | - | 1 | 129 | |||
AK141615.1 | - | 2, 3 | 307 | |||
BB622061.1 | - | 2, 3 | 367 | |||
BC021479.1 | - | 511 | 5344198 | |||
BC051144.1 | - | 2, 3 | 279 | 6588980 | ||
BC085302.1 | - | 2, 3 | 254 |
