siRNA Details
Gene Symbol: Bsph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080942.2 | NP_001074411.1 | 3 | 165 | ||
GenBank mRNA | DQ227499.1 | - | 3 | 162 | ||
DQ664198.1 | - | 3 | 166 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bsph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080942.2 | NP_001074411.1 | 3 | 165 | ||
GenBank mRNA | DQ227499.1 | - | 3 | 162 | ||
DQ664198.1 | - | 3 | 166 |