siRNA Details
Gene Symbol: Bsph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080942.2 | NP_001074411.1 | 1 | 31 | ||
GenBank mRNA | AK020324.1 | - | 1 | 32 | ||
DQ227499.1 | - | 1 | 28 | |||
DQ664198.1 | - | 1 | 32 |
