siRNA Details
Gene Symbol: Adamtsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029967.3 | NP_084243.3 | 5 | 729 | ||
GenBank mRNA | AK045085.1 | - | 5 | 625 | ||
BC062814.1 | - | 5 | 665 | |||
BC139339.1 | - | 5 | 608 | |||
BC139367.1 | - | 5 | 608 | |||
CV555842.1 | - | 1 | 71 |
