siRNA Details
Gene Symbol: 6030458C11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166360.1 | NP_001159832.1 | 2 | 235 | ||
NM_029998.3 | NP_084274.1 | 2 | 235 | |||
GenBank mRNA | AK020074.1 | - | 2 | 224 | ||
AK035859.1 | - | 2 | 220 | |||
AK036721.1 | - | 2 | 125 | |||
AK038561.1 | - | 2 | 220 | |||
AK054133.1 | - | 2 | 209 | |||
AK078182.1 | - | 2 | 220 | |||
AK081625.1 | - | 2 | 201 | |||
AK144748.1 | - | 2 | 188 | |||
AK152195.1 | - | 2 | 228 | |||
AK162187.1 | - | 2 | 249 | |||
AK163412.1 | - | 2 | 215 | |||
BC028769.1 | - | 2 | 191 | |||
BC099607.1 | - | 2 | 191 | |||
BC119058.1 | - | 2 | 170 | 8733807 | ||
BC137759.1 | - | 2 | 170 | 8860778 |
