siRNA Details
Gene Symbol: Crisp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030033.1 | NP_084309.1 | 4 | 408 | ||
GenBank mRNA | AK020340.1 | - | 5 | 469 | ||
AY705449.1 | - | 4 | 408 | |||
DQ200908.1 | - | 6 | 585 |
