siRNA Details
Gene Symbol: Klhdc8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030075.2 | NP_084351.1 | 9 | 1865 | ||
GenBank mRNA | AK019849.1 | - | 9 | 1863 | ||
AK154428.1 | - | 9 | 1966 | |||
AK215637.1 | - | 1 | 29 | |||
BC024793.1 | - | 2 | 1270 | |||
BC048918.1 | - | 4 | 828 | 6309434 | ||
BC060089.1 | - | 2 | 2588 |
