siRNA Details
Gene Symbol: Klhdc8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030075.2 | NP_084351.1 | 10 | 2049 | ||
GenBank mRNA | AK019849.1 | - | 10 | 2047 | ||
AK154428.1 | - | 10 | 2150 | |||
BC024793.1 | - | 2 | 1593 | |||
BC048918.1 | - | 5 | 1012 | 6309434 | ||
BC060089.1 | - | 2 | 2911 |
