siRNA Details
Gene Symbol: Cypt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177380.1 | NP_001170851.1 | 1 | 110 | ||
GenBank mRNA | AK018908.1 | - | 1 | 110 | ||
BC100466.1 | - | 1 | 110 |
Gene Symbol: Cypt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191032.1 | NP_001177961.1 | 1 | 110 | ||
GenBank mRNA | BC099555.1 | - | 1 | 110 |
