siRNA Details
Gene Symbol: Cfap61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175280.3 | NP_780489.2 | 7 | 726 | ||
GenBank mRNA | AK019712.1 | - | 727 | |||
AK038772.1 | - | 7 | 727 | |||
AK086391.1 | - | 478 | ||||
BC107030.1 | - | 7 | 712 | 40050496 | ||
BC107031.1 | - | 7 | 707 | 40050497 |
